Specifications
- Antibody type:Primary
- Antigen name:Dyslexia susceptibility 2
- Antigen symbol:DYX2
- Clonality:Polyclonal
- Conjugation:Cy7®
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:DYX2|DLX2|Dyslexia-associated protein KIAA0319 DYX 2|DYLX 2|MGC176717.|DYLX2|K0319_HUMAN|Kiaa0319|Dyslexia susceptibility 2|DLX 2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
DYX2 is a 1072 amino acid single-pass transmembrane protein that contains one MANSC domain and two PKD (Polycystic Kidney Disease) domains, which are usually found in the extracellular regions of proteins and are involved in protein-protein interactions. In DYX2, it is likely that its PKD domains mediate the interaction between neurons and glial fibers during neuronal migration. When overexpressed, this plasma membrane protein colocalizes with EEA1 (early endosome antigen 1) in large intracellular vesicles, suggesting that it is endocytosed and recycled. DYX2 is highly expressed in brain cortex, cerebellum, amygdala, putamen and hippocampus. Defects in the gene encoding DYX2 may be the cause of dyslexia type 2, a relatively common disorder that is characterized by reading performance impairment in the absence of sensory or neurologic disability. There are three isoforms of DYX2 that are produced as a result of alternative splicing events
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: DYX2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat