Anti-INSL3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-11301R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Insulin-like 3 (Leydig cell)
- Antigen symbol:INSL3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:RLNL|ley-I-L|RLF
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This gene encodes a member of the insulin-like hormone superfamily. The encoded protein is mainly produced in gonadal tissues. Studies of the mouse counterpart suggest that this gene may be involved in the development of urogenital tract and female fertility. This protein may also act as a hormone to regulate growth and differentiation of gubernaculum, and thus mediating intra-abdominal testicular descent. Mutations in this gene may lead to cryptorchidism. Alternate splicing results in multiple transcript variants. [provided by RefSeq, May 2012].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: INSL3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human