Order Entry
Italy
ContactUsLinkComponent
 
Anti-PCDHGC3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-11159R-A488
Supplier:  Bioss
Anti-PCDHGC3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-11159R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proto-Cadherin gamma subfamily C3
  • Antigen symbol:
    PCDHGC3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    PCDH gamma C3|Protocadherin 43|Protocadherin 2|Protocadherin43|PC 43|PCDH2|PCDHGC 3|PC43|PCDH 2|Protocadherin gamma subfamily C 3|PCDGK_HUMAN.|Cadherin like 2|Protocadherin gamma C3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin gene clusters, designated alpha, beta and gamma, all of which contain multiple tandemly arranged genes. PCDH2 (protocadherin-2), also known as PCDHGC3 (protocadherin gamma subfamily C, 3) or PC43, is a 934 amino acid single-pass type I membrane protein that contains six cadherin domains and belongs to the protocadherin gamma family. Functioning as a calcium-dependent cell-adhesion protein, PCDH2 is thought to be involved in the establishment and maintenance of neuronal connections within the brain. Multiple isoforms of PCDH2 exist due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCDHGC3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat