Anti-NKB Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-0070R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Neurokinin B
- Antigen symbol:NKB
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:6866
- Antigen synonyms:NKB|PRO1155|ZNEUROK1|NKNB
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:81-94/121
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Tachykinins are active peptides which excite neurons, evoke behavioral responses, are potent vasodilators and secretagogues, and contract (directly or indirectly) many smooth muscles (By similarity). Is a critical central regulator of gonadal function.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NKB
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat