Anti-MEK Kinase-1 Rabbit Polyclonal Antibody [clone: W197]
Catalog # BWRLBS9126
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:MEK Kinase-1
- Clonality:Polyclonal
- Clone:W197
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 170-220 of Human MEK Kinase-1.
MEK Kinase-1 (W197) pAb detects endogenous levels of MEK Kinase-1 protein.
Type: Primary
Antigen: MEK Kinase-1
Clonality: Polyclonal
Clone: W197
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat