Anti-EMR4 Rabbit Polyclonal Antibody [clone: I140]
Catalog # BWRLBS2792
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:EMR4
- Clonality:Polyclonal
- Clone:I140
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 108-164 of Human EMR4.
EMR4 (I140) pAb detects endogenous levels of EMR4 protein.
Type: Primary
Antigen: EMR4
Clonality: Polyclonal
Clone: I140
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat