Anti-Rsk-1 Rabbit Polyclonal Antibody [clone: E567]
Catalog # BWRLBS1779
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Rsk-1
- Clonality:Polyclonal
- Clone:E567
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 536-586 of Human Rsk-1.
Rsk-1 (E567) pAb detects endogenous levels of Rsk-1 protein
Type: Primary
Antigen: Rsk-1
Clonality: Polyclonal
Clone: E567
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat