Anti-ErbB2/HER2 Rabbit Polyclonal Antibody [clone: K676]
Catalog # BWRLBS2463
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ErbB2/HER2
- Clonality:Polyclonal
- Clone:K676
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 641-690 of Human ErbB2/HER2.
ErbB2/HER2 (K676) pAb detects endogenous levels of ErbB2/HER2 protein.
Type: Primary
Antigen: ErbB2/HER2
Clonality: Polyclonal
Clone: K676
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat