Benchtop meter-attached electrode stand, Orion™ Star A series
Catalog # ORIOSTARA-BEA
Supplier: Thermo Orion
Specifications
- For:Orion Star A110 + A210 series, Orion Versa Star benchtop meters
- Description:Benchtop meter-attached electrode stand (electrode holder arm and head), Orion™ Star A series, 360° motion (without weighted base)
- Seq:103
- Environmentally Preferable:
- Pk:1
Specifications
About this item
This meter-attached electrode stand can be used to easily place sensors in samples.
- Simplifies rinsing and storage between samples
- Cable management system minimises electrode breakage
- 360° motion allows electrodes to be moved vertically, horizontally and front-to-back positioning into beakers or sample containers
Can be attached to either side of Thermo Scientific™ Orion™ Versa Star benchtop meters and Thermo Scientific™ Orion™ Star A210 and Star A110 series benchtop meters. Can also be used with weighted base for compatibility with any meter system (portable or benchtop).
Ordering information: The weighted base needs to be ordered separately.
Delivery: Includes meter bracket, arm and electrode holder.