Tubing for rotarus® peristaltic pumps, Tygon® Chemical
Supplier: HIRSCHMANN
Certificates
About this item
These tubes are used in chemical application for its very high chemical resistance to acids, bases, salts, ketones, alcohols.
- Temperature resistance from –60 to 74 °C
- Hardness (Shore durometer A): 61
Frequently Bought Together