Order Entry
Italy
Orders LinkContactUsLinkComponent
Hydraulic pellet press, KBr
Hydraulic pellet press, KBr
Hydraulic pellet press, 15 t
Catalog # PERC51004599
Supplier:   PerkinElmer
Hydraulic pellet press, KBr
Catalog # PERC51004599
Supplier:  PerkinElmer
Hydraulic pellet press, KBr
Hydraulic pellet press, 15 t
Price
7 230,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
This compact hydraulic press is used with the KBr die to prepare KBr pellets.
Pressure is produced by operating a pump lever.
Ordering information: Evacuable KBr die, KBr pellet holder and adjustable adapter have to be ordered separately.
Specifications
  • Description:
    Hydraulic pellet press, 15 t
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Potassium bromide pellets die, Ø = 13 mm
    Potassium bromide pellets die, Ø = 13 mm
    PERC51001144
    Log in for availability
    1 * 1 pezzi - 1 090,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Potassium bromide pellet holder
    Potassium bromide pellet holder
    PERC04810949
    Log in for availability
    1 * 1 pezzi - 337,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Adjustable adapter
    Adjustable adapter
    PERCL1060351
    Log in for availability
    1 * 1 pezzi - 125,00 €