Order Entry
Italy
ContactUsLinkComponent
Quantitative filter papers, ashless grades, grade 589/1 and 589/1 ½, Whatman™
Quantitative filter papers, ashless grades, grade 589/1 and 589/1 ½, Whatman™
Certificates
undefined
Quantitative filter papers, ashless grades, grade 589/1 and 589/1 ½, Whatman™
Certificates

Specifications

  • Catalog No:
  • 512-3605
  • 512-3604
  • 512-0650
  • 512-3603
  • 512-3602
  • 512-3601
  • Grade:
  • 589/1
  • 589/1
  • 589/1 ½
  • 589/1
  • 589/1
  • 589/1
  • Weight:
  • 80 g/m²
  • 80 g/m²
  • 80 g/m²
  • 80 g/m²
  • 80 g/m²
  • 80 g/m²
  • Thickness:
  • 190 µm
  • 190 µm
  • 190 µm
  • 190 µm
  • 190 µm
  • 190 µm
  • Particle retention:
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • 12 - 25 µm
  • Filtration speed:
  • 25 s (Herzberg)
  • 25 s (Herzberg)
  • 25 s (Herzberg)
  • 25 s (Herzberg)
  • 25 s (Herzberg)
  • 25 s (Herzberg)
  • Ash content:
  • 0,010%
  • 0,010%
  • 0,010%
  • 0,010%
  • 0,010%
  • 0,010%

Specifications

Frequently Bought Together