Myelin myelin basic protein
Catalog # BIRBORB79674-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Source:
- Species:Myelin
- Environmentally Preferable:
- Storage conditions:Store at -2 to -8 °C.
- Protein/peptide name:Myelin Basic Protein
- Purity:Greater than 95% pure SDS-PAGE. Protein G Chromatography