Order Entry
Italy
ContactUsLinkComponent
Anti-KRT7 Mouse Monoclonal Antibody (CF640R) [clone: KRT7/1198]
Anti-KRT7 Mouse Monoclonal Antibody (CF640R) [clone: KRT7/1198]
Supplier:  Biotium
Certificates
Anti-KRT7 Mouse Monoclonal Antibody (CF640R) [clone: KRT7/1198]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC401198-500
  • BNC401198-100
  • Antigen symbol:
  • KRT7
  • KRT7
  • Antigen name:
  • Cytokeratin 7
  • Cytokeratin 7
  • Antigen synonyms:
  • KRT7|Krt2-7|Sarcolectin|SCL|K2C7|Keratin 55K Type II Cytoskeletal|CK-7|Type-II Keratin Kb7|Type II Mesothelial Keratin K7|Keratin Simple Epithelial Type 1 K7|Keratin Type II Cytoskeletal 7
  • KRT7|Krt2-7|Sarcolectin|SCL|K2C7|Keratin 55K Type II Cytoskeletal|CK-7|Type-II Keratin Kb7|Type II Mesothelial Keratin K7|Keratin Simple Epithelial Type 1 K7|Keratin Type II Cytoskeletal 7
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone KRT7/1198
  • Clone KRT7/1198
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 411501
  • 411501
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together