About this item
PDCD-1 (programmed cell death-1 protein), also designated CD279, is a type I transmembrane receptor and a member of the immunoglobin gene superfamily. It is expressed on activated T-cells, B-cells, and myeloid cells. Anti-PDCD-1 is a marker of angioimmunoblastic lymphoma and suggests a unique cell of origin for this neoplasm. Unlike CD10 and BCL6, PDCD-1 is expressed by few B-cells, so anti-PDCD-1 may be a more specific and useful diagnostic marker in angioimmunoblastic lymphoma. In addition, PDCD-1 expression provides evidence that angioimmunoblastic lymphoma is a neoplasm derived from germinal center-associated T-cells.
Type: Primary
Antigen: PDCD1
Clonality: Monoclonal
Clone: PDCD1/922
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNCB0922-500
- BNCB0922-100
- Antigen symbol:
- PDCD1
- PDCD1
- Antigen name:
- Programmed cell death 1
- Programmed cell death 1
- Antigen synonyms:
- hPD-1|CD279|hSLE1|Systemic lupus erythematosus susceptibility 2|Protein PD-1|PDCD1|PD1|Programmed Cell Death Protein 1|SLEB2
- hPD-1|CD279|hSLE1|Systemic lupus erythematosus susceptibility 2|Protein PD-1|PDCD1|PD1|Programmed Cell Death Protein 1|SLEB2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- Clone PDCD1/922
- Clone PDCD1/922
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 158297
- 158297
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide