Anti-NFH Rabbit Polyclonal Antibody
Catalog # BOSSBS-10680R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:200 kD Neurofilament Heavy
- Antigen symbol:NFH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:4744
- Antigen synonyms:Neurofilament triplet H protein|KIAA0845|NF-H|NHC|NFH|Neurofilament heavy polypeptide|200 kDa neurofilament protein
- Storage buffer:Aqueous buffered solution containing 100 μg/ml BSA, 50% glycerol and 0,09% sodium azide. Store at −20 °C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4 °C
- Immunogen:40-75/1026
- Purification:Purified by Protein A
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Neurofilaments usually contain three intermediate filament proteins: L, M, and H which are involved in the maintenance of neuronal caliber. NF-H has an important function in mature axons that is not subserved by the two smaller NF proteins.
Recommended Dilutions: IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.
Type: Primary
Antigen: NFH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat