Order Entry
Italy
ContactUsLinkComponent
 
Restriction enzymes, AgeI, FastDigest™, Fermentas
  FD1464
 :  
undefined
Restriction enzymes, AgeI, FastDigest™, Fermentas
  FD1464
 :  

 

  • Pk:
    1 KIT
  • Application:
  • Compatible buffer:
    FastDigest Green Buffer or FastDigest Buffer (colorless)
  • Overhang type:
    5' overhang
  • Source:
  • Description:
    AgeI, FastDigest™
  • Environmentally Preferable:
  • Enzyme name:
    FastDigest BshTI
  • No. of reactions:
    20 (20 µl)

 

 

Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.

  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Designed to complete digestion in 5 to 15 minutes
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications