Order Entry
Italy
ContactUsLinkComponent
Anti-CD53 Mouse Monoclonal Antibody (CF640R) [clone: 161-2]
Anti-CD53 Mouse Monoclonal Antibody (CF640R) [clone: 161-2]
Supplier:  Biotium
Certificates
Anti-CD53 Mouse Monoclonal Antibody (CF640R) [clone: 161-2]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC400324-100
  • BNC400324-500
  • Antigen symbol:
  • CD53
  • CD53
  • Antigen synonyms:
  • Leukocyte surface antigen CD53|CD53 glycoprotein|CD53 tetraspan antigen|TSPAN25|Tetraspanin-25 (Tspan-25)|Cell surface glycoprotein CD53
  • Leukocyte surface antigen CD53|CD53 glycoprotein|CD53 tetraspan antigen|TSPAN25|Tetraspanin-25 (Tspan-25)|Cell surface glycoprotein CD53
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone 161-2
  • Clone 161-2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 443057
  • 443057
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together