Kanamycin sulphate in solution
Catalog # ICNA091672048
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:50 mL
- Seq:2370
- Environmentally Preferable:
Specifications
About this item
Synonyms: Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate
:
:582.6 g/mol
CAS Number:25389-94-0
Formula:C₁₈H₃₈N₄O₁₅S
MDL Number:MFCD00070253