Alcian blue 8GX, dark blue powder
Catalog # ICNA0215262425
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
- UV/VIS: λmax (water):620 - 630 nm
Specification Test Results
About this item
:
:1298.86 g/mol
Storage Temperature:Ambient
CAS Number:75881-23-1
Formula:C₅₆H₆₈Cl₄CuN₁₆S₄
MDL Number:MFCD00010720