Human angiopoietin-1 ELISA kit
Catalog # BIRBORB138056-96
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:96
- Host:
- Primary antibody reactivity:Human
- Description:Human Angiopoietin-1 ELISA Kit
- Environmentally Preferable:
- Detection range:156 - 10000 pg/ml
- Sensitivity:<10 pg/ml
Specifications
About this item
Human Angiopoietin-1 ELISA Kit is used for quantitative detection of human Angiopoietin-1 in cell culture supernates, serum, plasma (heparin, EDTA), saliva and human milk.
Angiopoietin 1, also called ANG1 is a type of angiopoietin and is encoded by the gene ANGPT1. Angiopoietins are proteins with important roles in vascular development and angiogenesis. The protein contributes to blood vessel maturation and stability, and may be involved in early development of the heart.