Chitosan (from Crustacean chitin), off-white powder
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Deacetylated chitin, Poly(D-glucosamine)
CAS Number:9012-76-4
:MFCD00149018
MDL Number:MFCD00161512
Density:0.15…0.3 g/cm³ (bulk density)
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
- Source:Crustacean Chitin
- Moisture:≤10%
Specification Test Results
Frequently Bought Together