Order Entry
Italy
ContactUsLinkComponent
Cryopreservation media, Cellvation, MP Biomedicals
Cryopreservation media, Cellvation, MP Biomedicals
Catalog # ICNA0920300M2
Supplier:  MP Biomedicals
CAS Number:  
Cryopreservation media, Cellvation, MP Biomedicals
Catalog # ICNA0920300M2
Supplier:  MP Biomedicals
CAS Number:  
Specifications
  • Pk:
    60 mL
  • Description:
    Cellvation cryopreservation media
  • Seq:
    02
  • Environmentally Preferable:
Specifications
About this item
Cellvation is a cryopreservation medium designed for optimal cell recovery and convenience.
  • Optimal cell recovery and convenience
  • No DMSO or serum
  • Animal component free
  • Preserve stem cells, hepatocytes, and tissue samples.
Cellvation does not contain DMSO or serum, and is scientifically engineered for serum-free cultures, as well as being ideally suited for all types of serum. Cellvation cryopreservation media is animal-component-free and has been shown to preserve the potential of stem cells for differentiation. In addition, the medium is recommended for the preservation of hepatocytes, tissue samples, and a wide variety of cell types and established cell lines.