Order Entry
Italy
ContactUsLinkComponent
 
Avantor® Seradigm Foetal Bovine Serum (FBS), Ultra-Low IgG, US Origin
 
undefined
Avantor® Seradigm Foetal Bovine Serum (FBS), Ultra-Low IgG, US Origin
 

 

  • Catalog No:
  • 10018-830
  • 10836-698
  • 10836-696
  • 10018-826
  • Endotoxin:
  • ≤5 EU/ml
  • ≤5 EU/ml
  • ≤5 EU/ml
  • ≤5 EU/ml
  • Haemoglobin:
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • Storage temperature:
  • Frozen
  • Frozen
  • Frozen
  • Frozen

 

Frequently Bought Together