Anti-CD24 Mouse Monoclonal Antibody (APC) [clone: SN3]
Catalog # ABNOMAB13851
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:CD24
- Clonality:Monoclonal
- Clone:SN3
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:100133941
- Size:100 reactions
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human CD24.
Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: SN3
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human