Anti-CHI3L1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB29938
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Chitinase 3-like 1
- Antigen symbol:CHI3L1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Virus,Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:1116
- Sequence:LDFISIMTYDFHGAWRGTTGHHSPLFRGQEDASPDRFSNTDYAVGYMLRL
- Size:100 μL
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of human CHI3L1.
Type: Primary
Antigen: CHI3L1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,Viruses