Sampling pouches, 400 series Kapak®
Supplier: ProAmpac
Certificates
About this item
63 micron high barrier, crystal clear, three side seal, heat sealable polyester or foil pouches.
- Pouches are microwaveable, boilable and freezable
- High barrier leak proof pouches help eliminate cross contamination of dangerous substances
- With 9 mm seals and tear notches
- Infrared and gamma ray resistant
Typical applications: Medical, industrial, food preservation, veterinarian, health and beauty, agriculture.
Frequently Bought Together