Disodium hydrogen phosphate dodecahydrate 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: sec-Sodium phosphate, Sodium hydrogen phosphate dodecahydrate, Disodium phosphate, Dibasic sodium phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, di-Sodium hydrogen phosphate dodecahydrate, Sodium monohydrogen phosphate dodecahydrate, di-Sodium hydrogen orthophosphate dodecahydrate
CAS Number:10039-32-4
:
Formula:Na₂HPO₄·12H₂O
MDL Number:MFCD00149181
Molecular Weight:358.14 g/mol
Melting Pt:35 °C
Merck Index:13,08733
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystals
- Water:57 to 61 %
- pH:8.4 to 9.6
- Titration with HClO4:≥98.5 %
- Heavy metals (as Pb):≤10 ppm
Specification Test Results
Frequently Bought Together