2-Amino-4-chloropyridine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-Chloro-2-pyridylamine, 4-Chloropyridin-2-amine
CAS Number:19798-80-2
:
Formula:C₅H₅ClN₂
MDL Number:MFCD04113820
Molecular Weight:128.56 g/mol
Melting Pt:130…132 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light brown Solid
- Infrared spectrum:Conforms
- Melting point:127 to 134 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together