Order Entry
Italy
ContactUsLinkComponent
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99.5%, AcroSeal®, purified by redistillation
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99.5%, AcroSeal®, purified by redistillation
Catalog # 433831000.
CAS Number:  
undefined
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99.5%, AcroSeal®, purified by redistillation
Catalog # 433831000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4170 to 1.4190 (20°C, 589 nm)
  • GC:
    ≥99.4 %
  • Color scale:
    ≤20 APHA

Specification Test Results

About this item

Synonyms: 1,2-Bis(dimethylamino)ethane, TMEDA, N,N,N',N'-Tetramethylethylenediamine
CAS Number:110-18-9
 :
Formula:C₆H₁₆N₂
MDL Number:MFCD00008335
 :116.21 g/mol
Boiling Pt:120…122 °C (760 mmHg)
Melting Pt:–55 °C
Density:0.77 g/cm³
UN:2372
 :GHS07
 :GHS05
 :GHS02
Flash Pt:17 °C
Storage Temperature:Ambient
ADR:3 II