Order Entry
Italy
ContactUsLinkComponent
 
3-[(Benzyloxycarbonyl)amino]propionaldehyde 95%
Catalog # ACRO427720010
CAS Number:  
undefined
3-[(Benzyloxycarbonyl)amino]propionaldehyde 95%
Catalog # ACRO427720010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to yellow Powder or solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    52 to 61 °C
  • GC:
    ≥94 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

About this item

Synonyms: 3-[(Benzyloxycarbonyl)amino]-1-propanal, Benzyl 3-oxopropylcarbamate, N-Phenylmethoxycarbonyl-3-aminopropanal
 :
 :207.229 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:65564-05-8
Formula:C₁₁H₁₃NO₃
MDL Number:MFCD05664298
Melting Pt:52…57 °C