N-Methyl-tert-butylamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14610-37-8
:
Formula:C₅H₁₃N
MDL Number:MFCD00042853
Boiling Pt:67…69 °C (760 mmHg)
Density:0.727 g/cm³
Molecular Weight:87.166 g/mol
Flash Pt:–26 °C
UN:2945
ADR:3 II
:GHS05
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3950 to 1.3970 (20°C, 589 nm)
- GC:96.0 %
Specification Test Results
Frequently Bought Together