Polyvinylacetate, M.W. ∼100000
Catalog # ACRO183265000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear Beads
- Infrared spectrum:Conforms
- Loss on drying:≤5 %
- GC:≥97.5 %
Specification Test Results
About this item
Synonyms: PVA
:
Storage Temperature:Ambient
CAS Number:9003-20-7
Formula:[CH₂CH(O₂CCH₃)]n
MDL Number:MFCD00084457
Density:1.18 g/cm³