Order Entry
Italy
ContactUsLinkComponent
 
Anti-FLAG Tag Rabbit Polyclonal Antibody (CF594)
  20434.
 :  Biotium
Anti-FLAG Tag Rabbit Polyclonal Antibody (CF594)
  20434.
 :  Biotium

 

  • Antibody type:
    Tag
  • Antigen name:
    FLAG Tag (DYKDDDDK)
  • Antigen symbol:
    FLAG Tag
  • Clonality:
    Polyclonal
  • Conjugation:
    CF® 594
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • Storage temperature:
    Store at –20 °C.
  • Concentration:
    1 mg/mL
  • Purification:
    Affinity purified
  • Size:
    100 μL
  • Pk:
    100 µl

 

 

Polyclonal anti-FLAG (DYKDDDDK) reacts with FLAG tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. This conjugate is prepared by labeling polyclonal rabbit anti-FLAG IgG (H+L) with CF™594 dye. CF™594 (Ex/Em 593/614 nm) is a deep-red dye spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugates due to its high quantum yield and exceptional water solubility. CF™594 also has excellent photostability ideal for demanding applications, such as confocal microscopy and single molecule imaging.

Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.

Type: Tag
Antigen: FLAG Tag
Clonality: Polyclonal
Clone:
Conjugation: CF594
Epitope:
Host: Rabbit
Isotype:
Reactivity: FLAG tag