Anti-Collagen I Antibody
Catalog # BSBTPB9938
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:collagen, type I, alpha 1
- Antigen symbol:COL1A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1277
- Antigen synonyms:EDSC|OI4|OI2|OI3|OI1
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Collagen alpha-1(I) chain(COL1A1) detection. Tested with WB, IHC-P in Human.
Type: Primary
Antigen: COL1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human