Anti-Bcl-X Antibody
BSBTPB9917
- Antibody type:Primary
- Antigen name:BCL2-like 1
- Antigen symbol:BCL2L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:598
- Antigen synonyms:BCLXL|BCLX|BCLXS|BCL2L|BCL-XL/S|Bcl-X|PPP1R52|bcl-xS|bcl-xL
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Bcl-2-like protein 1(BCL2L1) detection. Tested with WB in Human.
Type: Primary
Antigen: BCL2L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human