Order Entry
Italy
ContactUsLinkComponent
Substrate for western blotting, NBT (nitro-blue tetrazolium chloride)
Substrate for western blotting, NBT (nitro-blue tetrazolium chloride)
Catalog # PIER34035
CAS Number:  
undefined
Substrate for western blotting, NBT (nitro-blue tetrazolium chloride)
Catalog # PIER34035
CAS Number:  
Specifications
  • Pk:
    1 g
  • Description:
    NBT (powder)
  • Seq:
    01
  • Environmentally Preferable:
Specifications
About this item
NBT (nitro-blue tetrazolium chloride) yields a blue-purple, water-insoluble product upon reduction of the tetrazolium salt. The product of reduction is NBT-formazan. This substrate is widely used for immunochemistry applications because the color is linear and stable. NBT is used as the substrate for both glucose oxidase and alkaline phosphatase systems.