Paraformaldehyde 4% in PBS
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Polyoxymethylene
CAS Number:30525-89-4
:
Formula:(CH₂O)n
MDL Number:MFCD00133991
Melting Pt:123
Flash Pt:70
Merck Index:13,07096
:GHS05
:GHS08
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together