Magnesium hydroxide carbonate tetrahydrate (∼40 - 43,5% MgO), Reagent Grade
: Magnesium carbonate basic, Magnesium carbonate hydroxide, Magnesium carbonate hydrated basic
:235-192-7
:
:4MgCO₃·Mg(OH)₂·4H₂O
:MFCD00149783
:2.16 g/cm³ (25 °C)
:467.67 g/mol (395.58 anhy)
:> 2500 °C
:14,5660
:Ambient
- Environmentally Preferable:
Frequently Bought Together