Anti-PSMA3 Rabbit Polyclonal Antibody
BOSSBS-9357R
: Bioss
- Antibody type:Primary
- Antigen name:Proteasome 20S Alpha 3
- Antigen symbol:PSMA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:5684
- Antigen synonyms:proteasome subunit C8|PSC3|macropain subunit C8|multicatalytic endopeptidase complex subunit C8|proteasome subunit alpha type-3|HC8|proteasome component C8
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
The proteasome is a multicatalytic proteinase complex with a highly ordered ring shaped 20S core structure. The core structure is composed of 4 rings of 28 non identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin dependent process in a non lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: PSMA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat