- Antibody type:Primary
- Antigen name:E1A binding protein p400
- Antigen symbol:EP400
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Form:liquid
- Gene ID:57634
- Antigen synonyms:TNRC12|CAGH32|P400
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
NuAR is a multi-protein histone acetyltransferase complex that functions to acetylate the nucleosomal Histones H4 and H2A, thereby activating transcription of select target genes. p400, also known as EP400 (E1A binding protein p400) or mDomino, localizes to the nucleus and is a component of the NuAR complex. Expressed in brain, liver, thymus, lung, spleen, colon and kidney, p400 regulates the transcriptional activity of proteins such as MZF-1 and contributes to the ATPase and helicase activities of NuA4. p400 is a SWI2/ SNF2-related protein that can interact with the adenovirus oncoprotein E1A, thus activating the proapoptotic activity of E1A. The ability of p400 to regulate transcriptional and apoptotic activity suggests that the NuAR complex may be a crucial component of cell proliferation, transformation and, possibly, carcinogenesis. Five isoforms of p400 exist due to alternative splicing events.
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: EP400
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat