Order Entry
Italy
ContactUsLinkComponent
 
Anti-FMNL1 Rabbit Polyclonal Antibody
  BOSSBS-6812R
 :  Bioss
Anti-FMNL1 Rabbit Polyclonal Antibody
  BOSSBS-6812R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Formin L1
  • Antigen symbol:
    FMNL1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Gene ID:
    752
  • Antigen synonyms:
    KW-13|FRL1|C17orf1B|FHOD4|CLL-associated antigen KW-13|Formin-like protein 1|C17orf1|FMNL
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

FMNL1 (Formin-like protein 1) gene encodes a formin-related protein. Formin-related proteins have been implicated in morphogenesis, cyokinesis, and cell polarity. An alternative splice variant has been described but the full length sequence has not yet been determined. FMNL1 possibly has a role in the control of cell motility and survival of macrophages.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: FMNL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat