Specifications
- Antibody type:Primary
- Antigen name:GDP Mannose Pyrophosphorylase B
- Antigen symbol:GMPPB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:29925
- Antigen synonyms:GTP-mannose-1-phosphate guanylyltransferase beta|KIAA1851|GMPPB|Mannose-1-phosphate guanyltransferase beta.|GMPPB_HUMAN|GDP mannose pyrophosphorylase B|GDP-mannose pyrophosphorylase B|Mannose 1 phosphate guanylyltransferase
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
GMPPB is a 360 amino acid protein that belongs to the transferase hexapeptide repeat family and is involved in protein modification pathways. Functioning as a GDP-mannose pyrophosphorylase, GMPPB enzymatically catalyzes the conversion of mannose-1-phosphate and GTP to GDP-mannose and a free phosphate, a reaction that is involved in the production of N-linked oligosaccharides. Defects in the gene encoding GMPPB that cause errors in the glycosylation pathway may lead to congenital disorders of glycosylation (CDG). CDGs are multisystemic diseases that often involve both the central and peripheral nervous systems and are often characterized by endocrine and coagulation disorders. GMPPB is expressed as two isoforms due to alternative splicing events.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: GMPPB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat