- Antibody type:Primary
- Antigen name:Ferredoxin reductase
- Antigen symbol:FDXR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:2232
- Antigen synonyms:ADXR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Adrenodoxin Reductase (ADX Reductase) is a mitochondrial flavoprotein that receives electrons from NADPH and thereby initiates the electron-transport chain serving mitochondrial cytochromes P450. ADX Reductase participates in cholesterol side chain cleavage in all steroidogenic tissues, steroid 11-? hydroxylation in the adrenal cortex, 25-OH-vitamin D3-24 hydroxylation in the kidney and sterol C-27 hydroxylation in the liver. Alternate splicing of ADX Reductase produces two isoforms. Human ADX Reductase maps to human chromosome 17q24-q25.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: Ferredoxin Reductase/ADXR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat