Order Entry
Italy
ContactUsLinkComponent
Anti-FAM134B Rabbit Polyclonal Antibody
Anti-FAM134B Rabbit Polyclonal Antibody
  BOSSBS-13136R
 :  Bioss
undefined
Anti-FAM134B Rabbit Polyclonal Antibody
  BOSSBS-13136R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Family with sequence similarity 134 member B
  • Antigen symbol:
    FAM134B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Gene ID:
    54463
  • Antigen synonyms:
    JK1|family with sequence similarity 134 member B|JK 1|FAM134B|Protein FAM134B.|HSAN2B|F134B_HUMAN
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

Required for long-term survival of nociceptive and autonomic ganglion neurons.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: FAM134B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat