Anti-CaMK2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-0541R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Calcium/calmodulin Dependent Protein Kinase 2
- Antigen symbol:CAMK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:815 816 817 818
- Antigen synonyms:CAMKD|CaM-kinase II delta chain|CaM-kinase II gamma chain|CaMK2 delta|CAMKB|CaMKII beta subunit|CaMK II subunit beta|CaM kinase II gamma subunit|MGC29528|CAM2|CAMK2D|CAMK2G|CAMKG|CAMK2N2|Calcium/calmodulin dependent protein kinase II beta|CaM kinase II delta subunit|kinase CaMK2-beta.|CAMK 2B|KCC2B|CAMK 2|CAMK2B|CAMK|Calcium/calmodulin dependent protein kinase IIB|Calcium/calmodulin dependent protein kinase type II beta chain|CAMK2|CAM 2|CaM kinase II beta chain|CaM kinase II beta subunit|CAMK-II|CaM kinase II|CaMK2 gamma|Calcium/calmodulin dependent protein kinase CaM kinase II beta|CaMK2 beta|CaMKIIB|CaMK II beta subunit|Calcium/calmodulin-dependent protein|Proline rich calmodulin dependent protein kinase
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:20-70/479
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
CaM-kinase II (CAMK2) is a prominent kinase in the central nervous system that may function in long-term potentiation and neurotransmitter release. Member of the NMDAR signaling complex in excitatory synapses it may regulate NMDAR-dependent potentiation of the AMPAR and synaptic plasticity
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: CaMK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat