Superdex 200 Increase small-scale SEC columns
Supplier: Cytiva
Certificates
About this item
Superdex 200 Increase size exclusion chromatography columns are well suited for high-resolution analysis and small-scale purification of monoclonal antibodies (mAb) and other biomolecules with Mr~ 10000 to 600000.
- Enhanced performance: Improved resolution and runtime compared to predecessor Superdex 200
- Very high resolution: Small bead size and narrow particle size distribution provide high resolution, for high protein purity
- High flow rates: Rigid beads give excellent pressure/flow properties
Superdex 200 Increase is a next-generation, dextran-agarose composite matrix for SEC. With smaller and more rigid beads than their predecessors, Superdex Increase columns deliver higher resolution purification in shorter run times.
Specifications
- Catalog No:
- 28-9909-46
- 28-9909-45
- 28-9909-44
- Max. pressure:
- 30 bar
- 30 bar
- 30 bar
- pH range:
- 3 - 12
- 3 - 12
- 3 - 12
- Storage:
- 4 to 30 °C, 20% Ethanol
- 4 to 30 °C, 20% Ethanol
- 4 to 30 °C, 20% Ethanol
Specifications
Related Information
Frequently Bought Together