Anti-KIN Antibody
Catalog # BSBTPB9607
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Kin17 DNA and RNA binding protein
- Antigen symbol:KIN
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:22944
- Antigen synonyms:BTCD|Rts2|KIN17
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for DNA/RNA-binding protein KIN17(KIN) detection. Tested with WB, IHC-P in Human;Mouse.
Type: Primary
Antigen: KIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse