Anti-EED Antibody
Catalog # BSBTPB9581
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:embryonic ectoderm development
- Antigen symbol:EED
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8726
- Antigen synonyms:HEED|WAIT1
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Polycomb protein EED(EED) detection. Tested with WB in Human;Rat.
Type: Primary
Antigen: EED
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat