Anti-SPEG Antibody Pair
Catalog # ABNOH00010290-AP11
Supplier: Abnova
Specifications
- Antigen name:SPEG Complex locus
- Antigen symbol:SPEG
- Host:Multiple
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:10290
- Antigen synonyms:CNM5|APEG1|APEG-1|BPEG|SPEGbeta|SPEGalpha
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human SPEG.
Antibody pair set content: 1. Capture Antibody: Rabbit affinity purified Polyclonal Anti-SPEG (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-SPEG, IgG1 kappa (20 µg);
Type:
Antigen: SPEG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human